missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97+%, extra pure

Product Code. 10092960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092960 100 g Glass bottle 100g
10246320 5 g Glass bottle 5g
10480681 500 g Glass bottle 500g
10350544 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10092960 Supplier Thermo Scientific Acros Supplier No. 217531000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Color Brown to Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 97+%
Linear Formula PbO2
Fieser 01,5331; 02,233; 03,168; 11,282
Merck Index 15, 5462
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol
IUPAC Name plumbanedione
Formula Weight 239.19
Percent Purity ≥97%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Lead(IV) oxide, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.