missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LDPE Lid for Rectangular Container

Product Code. 10125903 Shop All Bürkle Products
Click to view available options
For Use With (Equipment):
48 L Container
50 L Container
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10125903

Brand: Buerkle™ 34400481

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Buerkle™ LDPE Lid for Rectangular Container is suitable for use with Buerkle™ Rectangular Containers.

  • Linear LDPE
  • Translucent
  • For use with rupture proof rectangular containers
TRUSTED_SUSTAINABILITY

Specifications

Product Type Container Lid
Material LDPE
Shape Rectangular
Closure Type Lid
Color Transparent
For Use With (Equipment) 48 L Container
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.