missing translation for 'onlineSavingsMsg'
Learn More

LDN 27219, Tocris Bioscience™

Product Code. 30001661
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001661 50 mg 50mg
30001643 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001661 Supplier Tocris Bioscience Supplier No. 4602/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Transglutaminase 2 (TG2) inhibitor; reversible

Transglutaminase 2 (TG2) inhibitor (IC50 = 0.25 μM). Binds at GTPase site; reversible.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312946-37-5
Molecular Formula C20H16N4O2S2
Molecular Weight (g/mol) 408.494
InChI Key WLBUICQBNZXIDJ-UHFFFAOYSA-N
Synonym 2-4-oxo-3,5-diphenylthieno 2,3-d pyrimidin-2-yl sulfanyl acetohydrazide, 2-4-oxo-3,5-diphenylthieno 2,3-d pyrimidin-2-yl sulfanylacetohydrazide, 4-oxo-3,5-diphenyl-3,4-dihydro-thieno 2,3-d pyrimidin-2-ylsulfanyl-acetic acid, 4-oxo-3,5-diphenyl-3,4-dihydro-thieno 2,3-d pyrimidin-2-ylsulfanyl-acetic acid hydrazide, 2-3,4-dihydro-4-oxo-3,5-diphenylthieno 2,3-d pyrimidin-2-yl thio-acetic acid hydrazide, 2-3,4-dihydro-4-oxo-3,5-diphenylthieno 2,3-d pyrimidin-2-yl thio acetic acid hydrazide, 2-4-oxo-3,5-diphenyl-3,4-dihydrothieno 2,3-d pyrimidin-2-yl sulfanyl acetohydrazide
PubChem CID 1150683
IUPAC Name 2-(4-oxo-3,5-diphenylthieno[2,3-d]pyrimidin-2-yl)sulfanylacetohydrazide
SMILES C1=CC=C(C=C1)C2=CSC3=C2C(=O)N(C(=N3)SCC(=O)NN)C4=CC=CC=C4

Specifications

Quantity 50 mg
Formula Weight 408.5
Percent Purity >99%
Chemical Name or Material LDN 27219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.