missing translation for 'onlineSavingsMsg'
Learn More

LDN-27219, MedChemExpress

Product Code. 16433167
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433167 50 mg 50mg
16427138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16433167 Leverantör Medchem Express Leverantörsnummer HY16693

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress LDN-27219 is a potent inhibitor of hTGase(Tissue transglutaminase) with an IC50 of 0.6 uM.

LDN-27219 is a potent inhibitor of hTGase(Tissue transglutaminase) with an IC50 of 0.6 uM.

Specifikationer

Chemical Name or Material LDN-27219
Physical Form Solid
CAS 312946-37-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16N4O2S2
Solubility Information DMSO : ≥ 125 mg/mL (306.00 mM)
SMILES O=C1N(C2=CC=CC=C2)C(SCC(NN)=O)=NC3=C1C(C4=CC=CC=C4)=CS3
Molecular Weight (g/mol) 408.5
Formula Weight 408.5
Percent Purity 95.0%
Grade Research
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.