missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-PY20 Antibody

Product Code. 10607795
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10607795 25 μg Each
10141050 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10607795 Supplier Invitrogen™ Supplier No. PV3528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine kinases
Applications Kinase Assay
Concentration 0.54mg/mL
Conjugate Tb (Terbium)/Lanthanide Chelates
Description LanthaScreen™ Tb-PY20 Antibody, 25μg, 0.54mg/mL Concentration, Tyrosine Kinases Antigen, Tb (Terbium) Conjugate, 340/490nm Excitation, 520nm Emission, 10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline) Storage Buffer, Used for
Molecular Weight of Antigen 150kDa
Quantity 25 μg
Regulatory Status RUO
Research Discipline Target and Lead Identification and Validation
Primary or Secondary Primary
Packaging 1 Each
Content And Storage Terbium-labeled anti-PY20 antibody in HEPES buffered saline. Store at -20°C.
Assay Entry Cell-based terbium phospho-specific activity (stable)
Excitation/Emission 340/490, 520 nm
Shipping Condition Dry Ice
Readout End Point
Target Gene Tyrosine Kinases
Target Entry IGF-1R
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.