missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit

Product Code. 10175772
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175772 1 mg 1mg
11525046 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10175772 Supplier Thermo Scientific™ Supplier No. PV4452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit

  • A member of the leucine zipper family of DNA binding proteins
  • ATF2 (activating transcription factor 2) is phosphorylated at Thr71 by members of the JNK and p38 families of kinases
  • For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)/GFP (EmGFP)
Quantity 1 mg
Content And Storage Terbium-labeled anti-ATF2 [pThr71] antibody in HEPES buffered saline. Store at -20°C.
Antibody Molecular Weight 150 kDa
Format 384-well Plate
Excitation/Emission 340/490, 520
Reactivity Human
Shipping Condition Dry Ice
Readout End Point
Reactivity abrv Hu
Target Enzyme Serine/Threonine Kinases
Technique TR-FRET
Validated Kinases MAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.