missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-anti-ubiquitin-FK2 antibody

Product Code. 10165164
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10165164 25 μg Each
10470635 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10165164 Supplier Invitrogen™ Supplier No. PV4733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin.

The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications Biochemical Proteasome Assays, Cellular Lysate Proteasome Assays
Concentration 0.53mg/mL
Conjugate Tb (Terbium)/Lanthanide Chelates
Purification Method Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Proteasome Biology
Primary or Secondary Primary
Packaging 1 Each
Target Species Human
Content And Storage Terbium-labeled anti-ubiquitin-FK2 antibody in HEPES buffered saline.

Store at -20°C.
Form Liquid
Excitation/Emission 340/490, 520 nm
Reactivity Human
Shipping Condition Dry Ice
Readout End Point
Reactivity abrv Hu
Target Gene Ubiquitin
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.