missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-PRAS40 [pSer183] Antibody

Product Code. 10525304
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10525304 25 μg Each
10093614 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10525304 Supplier Thermo Scientific™ Supplier No. PV5578

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use in combination with the LanthaScreen™ PRAS40 HEK293E cell line (K1528) to evaluate inhibitors in medium to high throughput screening applications.

  • PRAS40 (Proline-rich AKT substrate, 40 kDa) is a cytosolic protein that is phosphorylated at position Ser183
  • It has been shown to bind to the mTORC1 complex and inhibit downstream signaling (i.e., phosphorylation of 4E-BP1 or p70S6K)
  • For use in combination with the LanthaScreen™ PRAS40 HEK293E cell line (K1528) to evaluate inhibitors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY

Spécification

Conjugate Tb (Terbium)/Lanthanide Chelates
Quantity 25 μg
Content And Storage Terbium-labeled pPRAS40 (pSer183) antibody in HEPES buffered saline. Store at -20°C.
Assay Entry Cell-based terbium phospho-specific activity (stable)
Excitation/Emission 340/90, 520
Reactivity Human
Shipping Condition Dry Ice
Readout End Point
Reactivity abrv Hu
Target Gene Ser/Thr Kinases
Target Entry PRAS40
Product Line LanthaScreen
Afficher plus Afficher moins

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.