missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ERK2 [pThr185/pTyr187] Antibody
Description
- A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2)
- For use in combination with the LanthaScreen™ ERK2 A375 cell line (K1798) to evaluate inhibitors in medium to high throughput screening applications
Specifications
Specifications
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
| Quantity | 25 μg |
| Content And Storage | Terbium-labeled pERK2 (pThr185/pTyr187) antibody in HEPES buffered saline. Store at -20°C. |
| Assay Entry | Cell-based terbium phospho-specific activity (stable) |
| Excitation/Emission | 340/490, 520 |
| Reactivity | Human |
| Shipping Condition | Dry Ice |
| Readout | End Point |
| Reactivity abrv | Hu |
| Target Gene | Ser/Thr Kinases, Tyrosine Kinases |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?