missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Biotin Antibody Kit
Description
The LanthaScreen™ terbium (Tb)-labeled anti-biotin antibody is a generic reagent that can be used to easily label biotinylated biomolecules for use in TR-FRET assays. Although the terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide, it can also be indirectly attached to a target molecule through a biotin-mediated association, in the form of a Tb-labeled streptavidin or anti-biotin antibody. Invitrogen’s Tb-labeled biotin-binding reagents are universal reagents that easily label biotinylated biomolecules with terbium for use in TR-FRET assays for proteases, kinases, etc.
Contents and Storage:
Each LanthaScreen™ Tb-labeled anti-biotin antibody is supplied in HEPES-buffered saline. The kit also includes 400μl of a fluorescein-labeled positive control and 100 ml of TR-FRET dilution buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C).
Contents and Storage:
Each LanthaScreen™ Tb-labeled anti-biotin antibody is supplied in HEPES-buffered saline. The kit also includes 400μl of a fluorescein-labeled positive control and 100 ml of TR-FRET dilution buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C).
Specifications
Specifications
| Applications | Kinase Assay, Enzyme and Protein Activity Assays |
| Concentration | 1.1mg/mL |
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
| Quantity | 25 μg |
| Regulatory Status | RUO |
| Research Discipline | TR-FET Assay Development |
| Primary or Secondary | Primary |
| Packaging | 1 Kit(s) |
| Includes | 0.1% Sodium azide |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?