missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-Biotin Antibody Kit

Product Code. 10135404
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10135404 1 mg Each
10627795 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10135404 Supplier Thermo Scientific™ Supplier No. PV4750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LanthaScreen™ Tb-anti-Biotin Antibody Kit

  • The LanthaScreen™ terbium (Tb)-labeled anti-biotin antibody is a generic reagent that can be used to easily label biotinylated biomolecules for use in TR-FRET assays
  • The terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide
  • It can also be indirectly attached to a target molecule through a biotin-mediated association, in the form of a Tb-labeled streptavidin or anti-biotin antibody
  • Invitrogen’s Tb-labeled biotin-binding reagents are universal reagents that easily label biotinylated biomolecules with terbium for use in TR-FRET assays for proteases, kinases, etc

Contents and Storage:

  • Each LanthaScreen™ Tb-labeled anti-biotin antibody is supplied in HEPES-buffered saline
  • The kit also includes 400 μL of a fluorescein-labeled positive control and 100 mL of TR-FRET dilution buffer
  • Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C)
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)/Lanthanide Chelates
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Dry Ice
Product Line LanthaScreen

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.