Learn More
Invitrogen™ LanthaScreen™ Eu-PY20 Antibody
Description
A general reagent to monitor tyrosine kinase activity, the PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with AlexaFluor 647-poly-GT (Cat. No. PV5693) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Conjugate | Europium/Lanthanide Chelates |
| Quantity | 1 mg |
| Content And Storage | Europium-labeled anti-PY20 antibody is supplied in HEPES-buffered saline. Store at -20°C. |
| Excitation/Emission | 340/665, 615, 340/615, 665 nm |
| Shipping Condition | Dry Ice |
| Readout | End Point |
| Target Gene | Tyrosine Kinases |
| Product Line | LanthaScreen |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.