missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Eu-anti-DYKDDDDK Antibody

Product Code. 10192928
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10192928 1 mg Each
10070808 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10192928 Supplier Thermo Scientific™ Supplier No. PV6027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LanthaScreen™ Eu-anti-DYKDDDDK Antibody

  • LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag)
  • This can be advantageous when working with partially purified protein
  • It is also useful in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
Conjugate Europium/Lanthanide Chelates
Quantity 1 mg
Content And Storage Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C.
Form Frozen
Excitation/Emission 340/615, 665
Shipping Condition Dry Ice
Product Line LanthaScreen

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.