missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Eu-anti-Biotin Antibody

Product Code. 10123078
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123078 25 μg Each
10214448 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10123078 Supplier Thermo Scientific™ Supplier No. PV5901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LanthaScreen™ Eu-anti-Biotin Antibody

A general reagent for use in assay development, the Europium-anti-biotin antibody binds to biotinylated peptide or physiological protein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg⁄ml
Conjugate Alexa Fluor 647/Europium/Lanthanide Chelates
Formulation Liquid, frozen
Quantity 25 μg
Regulatory Status RUO
Research Discipline Pharma and Biopharma
Primary or Secondary Primary
Packaging 1 Each
Includes 0.1% Sodium azide
Content And Storage Europium-labeled anti-Biotin antibody in HEPES buffered saline. Store at -20°C.
Form Frozen
Excitation/Emission 340⁄615, 665
Shipping Condition Dry Ice
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.