Learn More
Invitrogen™ Laminin gamma-1 Monoclonal Antibody (LN82-13)

Description
This antibody recognizes the N-terminal region of human laminin gamma 1 chain. This antibody does not inhibit cell adhesion. This antibody reacts with rabbit laminin. This antibody does not react with mouse laminin. Monoclonal antibody was obtained by fusing the mouse myeloma cell-line P3U1 with spleen cells of BALB/c mouse after immunization with human placenta laminin. Reconstitution: Dissolve the lyophilized antibody in 50 μL of distilled water (final concentration: 2.0 mg/mL). This solution can be used as a stock solution. If dilution is necessary for your application, dilute the stock solution with the following Dilution solution just prior to use. When the entire amount of antibody is to be used over a short time period, it may be dissolved directly in 500 mL or more of the Dilution solution. Be sure to store the antibody at a minimum concentration of 2.0 mg/mL. A lower antibody concentration may result in decreased stability. Reconstituted antibody solution should contain 0.1% sodium azide as a preservative when stored at 4°C. Storage : 4°C, This product does not contain preservative. The stock solution(2.0 mg/mL) should be stored in aliquots at -20°C for 1 year, or should be stored at 4°C for 6 months after adding 0.1% sodium azide. Avoid repeated freeze-thaw cycles. Application: Western blot analysis under non-reducing and non-heating condition (can not be used under reducing condition). Immunohistochemical detection of laminin on frozen or p...
Specifications
Specifications
| Antigen | Laminin gamma-1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | LN82-13 |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and no preservative; pH 7.4 |
| Gene | LAMC1 |
| Gene Accession No. | P11047 |
| Gene Alias | AA408497; B2e; basement membrane protein; LAMA; LAMA1; Lama-1; LAMB2; LAMC1; Laminin A chain; laminin B2 chain; laminin gamma 1 chain; laminin S; laminin subunit alpha 1; laminin subunit alpha-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit gamma; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit gamma; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit gamma; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.