missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Warning Safety Signs

Product Code. 10061394
Change view
Click to view available options
Width (Metric):
100 mm
200 mm
Displayed Text:
Danger Zone Warning Pictogram - !
Warning about skid risk
Warning of industrial trucks
Warning of tripping hazard
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Displayed Text unitSize
10061394 100 mm Warning of tripping hazard Each
10151984 100 mm Warning about skid risk Each
10041344 100 mm Danger Zone Warning Pictogram - ! Each
10264894 200 mm Danger Zone Warning Pictogram - ! Each
10441074 100 mm Warning of industrial trucks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10061394 Supplier Brady™ Supplier No. 250276
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance
Alternate Part Number PIC 325-TRI 100-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of tripping hazard
Product Type Warning Safety Sign
Color Black on Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.