Learn More
Lenz Laborglasintrumente Laboratory Bottles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07105094
This item is not returnable.
View return policy
Description
- Acc. to ISO 4796, with DIN thread
- Material: DURAN™
- With easy-to-read scale and large labelling field for easy marking
- With screw cap and pouring ring (material: PP, blue; temperature resistance: +140 °C)