missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ MicroDispo™ Disposable Sampler
GREENER_CHOICEBIOPHARMA

Product Code. 10093982
Change view
Click to view available options
Length (Metric):
500 mm
1000 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Sterility unitSize
10093982 1000 mm Sterile Pack of 20
10738513 500 mm Non-sterile Pack of 20
10554673 500 mm Sterile Pack of 20
10626013 1000 mm Non-sterile Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10093982 Supplier Buerkle™ Supplier No. 53932251
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Extracts small sample quantities for analysis. Buerkle™ SteriPlast™ MicroDispo takes precise target point samples of free-flowing powders and small-grain bulk materials.

  • Ergonomic handle
  • HDPE
  • Cleanroom manufactured
  • Optionally sterilized by gamma rays
  • 21 mm dia.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disposable Sampler
Material HDPE
Capacity (Metric) 10 mL
Disposable Single-use
Sterility Sterile
Length (Metric) 1000 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.