missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Potassium hydrogen tartrate, 99%

Product Code. 10196773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10196773 5 kg Plastic drum 5kg
10330594 250 g Plastic bottle 250g
10246420 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10196773 Supplier Thermo Scientific Acros Supplier No. 222670051
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
PubChem CID 24193652
IUPAC Name 2,3-dihydroxybutanedioic acid;potassium
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

Color Colorless or White
Density 1.9500g/mL
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic drum
Linear Formula KO2CCH(OH)CH(OH)CO2H
Quantity 5 kg
Beilstein 03, 529
Merck Index 15, 7736
Specific Gravity 1.95
Specific Rotation Condition + 32.50 (20.00°C c=10,1N NaOH)
Specific Rotation + 32.50
Formula Weight 188.18
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material L(+)-Potassium hydrogen tartrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.