missing translation for 'onlineSavingsMsg'
Learn More

L-Arabinitol, Mikromol™

Product Code. 17978134
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17978134 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 17978134

Brand: Mikromol MM1451.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Arabinitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-(-)-Arabitol
CAS 7643-75-6
Analyte or Component Names L-(-)-Arabitol
Shipping Condition Room Temperature
Molecular Formula C5 H12 O5
Certifications/Compliance ISO 17025
Synonym L-Arabinitol, (-)-Arabitol, L-(-)-Arabitol, L-Arabitol, L-Lyxitol, L-arabino-Pentitol, NSC 83253, Xylitol Imp. A (EP), Xylitol Impurity A
InChI Formula InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m0/s1
SMILES OC[C@H](O)C(O)[C@@H](O)CO
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 152.15
Quantity 100 mg
Formula Weight 152.0685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.