missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-2-Hydroxyglutaric acid, MedChemExpress
Description
L-2-Hydroxyglutaric acid is an epigenetic modifier and putative oncometabolite in renal cancer. L-2-Hydroxyglutaric acid can inhibit histone demethylases and hence promote histone methylation. L-2-Hydroxyglutaric acid inhibits mitochondrial creatine kinase (Mi-CK) activity with Km and Ki of 2.52 mM and 11.13 mM, respectively.
Specifications
Specifications
| Chemical Name or Material | L-2-Hydroxyglutaric acid |
| CAS | 13095-48-2 |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H8O5 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | (S)-2-Hydroxyglutaric acid |
| SMILES | O=C(O)[C@@H](O)CCC(O)=O |
| Molecular Weight (g/mol) | 148.11 |
| Formula Weight | 148.11 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?