missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures

Available in natural (clear) or choice of four colors

Brand:  DWK Life Sciences 73660-18

Product Code. 10135714

  • ONLINE EXCLUSIVE PRICE
    £126.00
    £110.81 / Pack of 1000
    Save £15.19
    12% Off
Only null left
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

Description

Description

Available in natural (clear) or choice of four colors

  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable

Caps for 38mm O.D. are designed for baffled culture flasks.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Closure
Polypropylene
Sterile
Single-use
Autoclavable
Natural
18 mm O.D. Plain-end Culture Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.