missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ MiniFix Micropipettor
Description
- Fully autoclavable
- Designed for fast, repetitive and precise operation
- Very compact in terms of size and weight
- Designed for routine or biological hazard work
- Kartell™ tips fit on the conical shaft as well as most other commercially available tips
- The plunger button has a different color based on dosing capacity
- Individually calibrated in compliance with ISO8655, the new international standard for piston-operated volumetric apparatus
- Carries the CE conformity mark
Specifications
Specifications
| Accuracy | ±0.3% |
| Certifications/Compliance | CE/IVD |
| Compatible Tips | Kartell™ 961 |
| Autoclavable | Autoclavable |
| Color | Red |
| Material | Polypropylene |
| Number of Channels | 1 |
| Type | Micropipette |
| Volume (Metric) | 500 μL |
| For Use With (Application) | Routine or biological hazard work, in situ analyses and sampling |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?