missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JNJ 0966

Description
JNJ 0966 is a pro-MMP9 activation inhibitor (IC50 = 440 nM). Inhibits the conversion of pro-MMP9 to active MMP9, without directly affecting MMP9 enzymatic activity. Exhibits selectivity for pro-MMP9 over pro-MMP1, 2 and 3. Inhibits cell invasion in in vitro assays. Reduces motor impairment in a mouse EAE model. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | N-[2-[(2-Methoxyphenyl)amino]-4'-methyl[4,5'-bithiazol]-2'-yl]acetamide |
| CAS | 315705-75-0 |
| Quantity | 50 mg |
| Molecular Formula | C16H16N4O2S2 |
| Formula Weight | 360.45 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?