missing translation for 'onlineSavingsMsg'
Learn More

isopropyl 2-bromobenzoate, Thermo Scientific™

Product Code. 10108834
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10108834 100 g 100g
10165892 1 g 1g
10045032 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10108834 Supplier Thermo Scientific Maybridge Supplier No. RDP00166FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59247-52-8
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
InChI Key QTCRCHHNKWSPHB-UHFFFAOYSA-N
PubChem CID 603261
IUPAC Name propan-2-yl 2-bromobenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1Br

Specifications

Assay Percent Range 97+%
Packaging Amber glass bottle
Quantity 100 g
Synonym isopropyl 2-bromobenzoate, isopropyl o-bromobenzoate, methylethyl 2-bromobenzoate, prop-2-yl 2-bromobenzoate, isopropyl 2-bromobenzoate #, isopropyl2-bromobenzoate, 2-bromobenzoic acid isopropyl ester, 2-bromobenzoic acid, isopropyl ester, benzoic acid, 2-bromo-,1-methylethyl ester, benzoic acid, 2-bromo-, 1-methylethyl ester
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material Isopropyl 2-bromobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.