missing translation for 'onlineSavingsMsg'
Learn More

Iso-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™

Product Code. 10192632 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192632 25 L Metal can 25 L
10365120 500 mL Amber glass bottle 500mL
10438210 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10192632 Supplier Fisher Chemical Supplier No. B/5150/25
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iso-Butyl Alcohol, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
MDL Number 4740

Specifications

Melting Point -108°C
Color Colorless
Boiling Point 108°C
Packaging Metal can
Molecular Formula C4H10O
Quantity 25 L
Vapor Density 2.6
Formula Weight 74.12g/mol
Vapor Pressure 11.7mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.