missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFN 530 Core Thermometer; Type K, J, T, E

Product Code. 10073722
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10073722 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10073722 Supplier Ebro Electronic™ Supplier No. 13405530A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Precise and very robust thermometer with a big display.

  • 2-channel high accuracy thermocouple thermometer
  • With Lemo connections
  • Robust design for years of industrial use
  • Approximately 5 year battery life 
  • Without probe

Core thermometer, precision, lithium battery, watertight, measurement, temperature, laboratory, sensor, display, robust, impact resistant, accuracy

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16; EN 13485
Disposable Single-use
For Use With (Application) For bakeries, butcheries and for the food industry
Height (Metric) 52 mm
Includes Thermometer, sensor, calibration certificate, instruction manual
Length (Metric) 335 mm
Material ABS Plastic
Model TFN 530
Resolution 0.1°C
Weight (Metric) 90 g
Product Type Precision Core Thermometer
Data Storage None
Description Without cable
Temperature Range (Metric) -50°C to +300°C or -50°C to +400°C
Calibrated Factory Calibrated
Battery Life Approx. 5 Years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.3°C
Immersion Length 120 mm
Display LCD
Width (Metric) 50 mm
IP Rating IP52
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.