missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViraPower™ Adenoviral Promoterless Gateway™ Expression Kit

Product Code. 10114512
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10114512 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10114512 Supplier Invitrogen™ Supplier No. K494000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

pAd/PL-DEST Gateway Vector , 293A Cell Line

Facilitate easy recombination-based cloning and adenoviral-based, transient expression of target gene in dividing and nondividing mammalian cells

  • Allows generation of adenovirus containing target gene where expression is driven by promoter of choice
  • Alternatively, vector may also be used to express small RNA molecules from their appropriate promoters
  • High efficiency and rapid recombination cloning
  • Produces high titer adenoviral stocks
  • Produces replication-incompetent virus for enhanced biosafety of system
  • Amenable for use in high-throughput applications
  • Human Ad5 sequences (ΔE3) and Viral Inverted Terminal Repeats (ITRs) for packaging of expression construct into virions
  • Ampicillin selection marker

Antibody and Cell Culture Production, Bioproduction, Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Adenoviral
Promoter None (Promoterless)
Product Type Adenoviral Expression Kit
Content And Storage • pAd/PL-DEST Gateway™ Vector (Store at -20°C)
• 293A Cell Line (Store in liquid nitrogen)
Protein Tag Untagged
Cloning Method Gateway
Quantity 1 kit
Vector pAd
Product Line Gateway, ViraPower
For Use With (Application) Clone your own Promoter, Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.