missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T-REx™ Jurkat Cell Line
Description
Includes
3 x 106 cells are supplied frozen in 1mL 90% complete medium and 10% DMSO.
- Stably express the tetracycline repressor protein
- T-REx cell lines are functionally tested by transient transfection with positive control vector pcDNA™4/TO/lacZ
- T-REx cell lines exhibit extremely low basal expression levels in repressed state and high expression upon induction with tetracycline
Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
Specifications
Specifications
| Content And Storage | 1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. |
| Cell Line | T-REx™ Jurkat Cell Line |
| Species | Human |
| Quantity | 1 x 107 cells |
| Product Line | T-REx |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?