Learn More
Invitrogen™ Quant-iT™ RNA Assay Kits and Quant-iT RNA HS Reagent
Achieve highly selective and accurate quantification of high-abundance RNA samples with the fluorescent Quant-iT RNA Reagent and Assay Kits. These kits are highly selective for RNA over double-stranded DNA, and provide sufficient material for at least 2000 assays in a 96-well microplate format.
Brand: Invitrogen™ Q33140
585.65 GBP valid until 2024-12-31
Use promo code "22910" to get your promotional price.
Code : 50
Additional Details : Weight : 0.38000kg
Description
Enable easy and accurate RNA quantitation with the Quant-iT RNA Assay Kit, which is also available in broad range (BR) and extended range (XR) configurations, as well as a stand-alone high sensitivity (HS) reagent. These Quant-iT RNA quantification kits are highly selective for RNA over dsDNA and can be used to generate fluorescence-based linear detection ranges of RNA.
Quant-iT RNA Reagent and Assay KitThe Quant-iT RNA Reagent and Assay Kit makes RNA quantification easy and accurate. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 5-100 ng of RNA.
Quant-iT RNA BR (Broad Range) Assay Kit
The Quant-iT RNA BR (Broad Range) Assay Kit provides an accurate and selective method for quantification of high-abundance RNA samples. While the original Quant-iT RNA kit has great sensitivity, the Quant-iT RNA Broad Range kit is ideal for microarray samples where the concentration of RNA is higher. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 20-1000 ng of RNA.
Quant-iT RNA XR (Extended Range) Assay Kit
The Quant-iT RNA XR (Extended Range) Assay Kit provides an accurate and selective method for the quantification of high-abundance RNA samples. The assay is highly selective for RNA and does not quantitate DNA, protein, or free nucleotides. The assay kit is designed to be accurate for initial RNA sample concentrations of 10 ng/μL to 10,000 ng/μL depending on sample volume, providing a linear detection range of 200–10,000 ng.
For each assay, simply dilute the reagent using the buffer provided, add the sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a fluorescence-based microplate reader. Common contaminants, such as salts, free nucleotides, solvents, detergents, and protein, are well tolerated.
Order Info
Shipping Condition: Room temperature
Specifications
RNA Quantitation | |
Microplate Reader | |
5 to 100 ng | |
Room Temperature |
Fluorescence | |
1000 Reactions (200 μL Assay Volume) | |
Quant-iT |