missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)
GREENER_CHOICE

Product Code. 10164582
Change view
Click to view available options
Assay:
dsDNA Quantitation, BR
dsDNA Quantitation, HS
Quantitation Range:
0.2 to 100 ng
4 to 1000 ng
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Assay Quantitation Range unitSize
10164582 dsDNA Quantitation, HS 0.2 to 100 ng 1 set
10555213 dsDNA Quantitation, BR 4 to 1000 ng 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10164582 Supplier Invitrogen™ Supplier No. Q33120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

For highly selective quantitation of double-stranded DNA over RNA, the Quant-iT dsDNA Assay Kits, high sensitivity and broad range, produce fluorescence signals that are linear in the ranges of 0.2-100 ng and 2-1000 ng of DNA, respectively.

Perform dsDNA quantification easily and quickly with Quant-iT dsDNA assay kits. Both the Quant-iT High-Sensitivity dsDNA Assay Kit and the Quant-iT Broad-Range dsDNA Assay Kit provide concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. These DNA assay kits are highly selective for double-stranded DNA over RNA, and, in the ranges of 0.2–100 ng DNA (for the HS dsDNA Assay Kit) and 4–1000 ng DNA (for the BR dsDNA Assay Kit), the fluorescence signal is linear.

The Quant-iT dsDNA High-Sensitivity Assay Kit and the Quant-iT dsDNA Broad-Range Assay Kit make DNA quantification easy and accurate. The kits provide concentrated assay reagent, dilution buffer, and prediluted DNA standards. Simply dilute the reagent 1:200, load 200 μL into the wells of a microplate, add 1–20 μL sample, mix, then read the fluorescence.

The assays are highly selective for double-stranded DNA over RNA, and, in the range of 0.2–100 ng for the HS assay or 4–1000 ng for the BR assay, the fluorescence signal is linear with DNA. The assays are performed at room temperature, and the signal is stable for three hours. Common contaminants such as salts, solvents, detergents, and protein are well tolerated in these assays.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Assay dsDNA Quantitation, HS
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
No. of Reactions 1000 Assays (200 μL assay volume)
Quantitation Range 0.2 to 100 ng
Product Line Quant-iT
Quantity 1 kit
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.