missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Pro 96 PCR Purification Kit

Product Code. 10768035
Change view
Click to view available options
Quantity:
4 plates kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10768035 4 plates kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10768035 Supplier Invitrogen™ Supplier No. K310096A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

Four 96-well plates and sufficient reagents to perform four 96-well purifications

The PureLink™ Pro 96 PCR Purification Kit combines advanced silica plate extraction chemistry with an optimized 96-well plate design for manual or automated processing of PCR cleanup in as little as 20 minutes.

The PureLink™ Pro 96 PCR Purification Kit combines advanced silica plate extraction chemistry with an optimized 96-well plate design for manual or automated processing of PCR cleanup in as little as 20 minutes. The PureLink™ Pro 96 PCR Purification Kits provide rapid and efficient removal of short primers, dNTPs, enzymes, short-failed PCR products, and salts from PCR products. The simple PureLink™ bind/wash/elute procedure facilitates routine purifications of double-stranded DNA fragments from 100bp to 12kb.

Improved plate design
The PureLink™ Pro 96 PCR Purification Kit provides optimized semi-skirted 96-well plates for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation, or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.

PCR products purified using the PureLink™ Pro 96 PCR Purification Kit demonstrate outstanding performance in downstream applications such as cloning automated fluorescent DNA sequencing, PCR, restriction mapping, and labeling.

High Throughput-Compatible

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The PureLink™ Pro 96 PCR Purification Kit includes 4 x 96-well plates, and sufficient reagents to perform 4 x 96-well purifications.

Store components at room temperature. All reagents are guaranteed stable for 12 months when properly stored.
Format Kit
Elution Volume Max 50 μL (centrifuge), 100 μL (vacuum), 150 μL
For Use With (Equipment) Biomek™ FX, Robotic Liquid Handling Systems, Vacuum Manifold, Eppendorf epMotion, Centrifuge, TECAN Freedom Evo, TECAN Genesis
High-throughput Compatibility High-throughput Compatible
Product Line PureLink
Product Type PCR Purification Kit
Purity or Quality Grade Molecular Biology Grade
Quantity 4 plates kit
Run Time 20 min.
Scale Mini
Shipping Condition Room Temperature
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Next-Generation Sequencing (NGS), Cloning
Final Product Type DNA
Isolation Technology Silica Plate
No. of Reactions 96 Preps
Sample Type PCR Products
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.