missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Growth Blocks

Product Code. 10033782
Change view
Click to view available options
Quantity:
50 Blocks
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10033782 50 Blocks Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10033782 Supplier Invitrogen™ Supplier No. 12256020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

Package of 50 blocks

PureLink Growth Blocks offer unique approach to high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System.

PureLink Growth Blocks offer unique approach to high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Package of 50 blocks The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.
Format 96 Deep-well Plate
For Use With (Application) Automated Nucleic Acid Purification, DNA Extraction
Product Line PureLink
Product Type Growth Block
Quantity 50 Blocks
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.