Learn More
Invitrogen™ pENTR™/TEV/D-TOPO™ Cloning Kit, with One Shot™ TOP10 Chemically Competent E. coli
Product Code. 10105082
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
Product Code. Quantity unitSize
10105082 20 Reactions Each
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10105082 Supplier Invitrogen™ Supplier No. K252520
For Research Use Only. All usage must comply with product instructions.
Includes
pENTR/TEV/D-TOPO vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot TOP10 Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid.
Use highly efficient, 5-minute cloning strategy to directionally clone blunt-end PCR product into vector
  • Blunt-end PCR products clone directionally into pENTR/TEV/D-TOPO entry vector at greater than 90% efficiency, and include TEV protease-dependent cleavage site for removal of N-terminal tags from expressed proteins
  • Rapidly shuttle cloned genes between multiple vector systems
  • Reliable performance
  • For access to Gateway System, PCR amplify gene of interest and add product straight to provided topoisomerase charged pENTR/SD/D-TOPO vector, incubate 5 minutes, and transform provided competent E. coli cells
  • Resulting attL containing Gateway Entry clones are ready for efficient recombination
  • Optimized pENTR/TEV/D-TOPO Vector includes Tobacco Etch Virus (TEV) recognition site for protease-dependent cleavage of N-terminal tag from recombinant protein
  • Vector has M13 and T7 primer sequencing sites and attL recombination sites flanking the PCR product insertion site, allowing clones to be easily sequence-verified and recombined into choice of vectors
  • Kanamycin resistance gene and pUC origin are used for selection and high-copy propagation in E. coli
  • Four base overhang on vector pairs with four base sequence designed into forward primer used in PCR reaction to provide directionality to topoisomerase ligation reaction
Cloning, Gateway Cloning
TRUSTED_SUSTAINABILITY
Specifications
Cloning Method Directional TOPO™
Content And Storage pENTR™⁄TEV⁄D-TOPO™ Cloning Kit contains pENTR⁄TEV⁄D-TOPO™ vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot™ TOP10 Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store Competent E. coli at -80°C. Store all other components at -20°C.
No. of Reactions 20 reactions
Product Line One Shot
Product Type TOPO Cloning Kit
Protein Tag N-Terminal
Quantity 20 Reactions
Vector pENTR