Learn More
Invitrogen™ E-Holder™ Platform
Description
This allows reproducible placement and loading from one gel to the next without having to make adjustments to the robot loading software. In addition, the E-Holder platform allows you to load samples on one E-Gel96 or E-Gel 48 gel while other gels are running in the mother or daughter E-Base combination.
- Gel Compatibility: E-Gel 48 and-Gel 96
- Gel Size: E-Gel 48 (13.5cm x 10.8cm) and E-Gel 96 (13.5cm x 10.8cm)
- Mode of Separation: Molecular Weight
The E-Holder platform can hold either E-Gel96, E-Gel48, or E-PAGE™ 96 Gels.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Each E-Holder™ platform is sold separately.
Specifications
Specifications
| Content And Storage | • Each E-Holder™ platform is sold separately Store at room temperature. |
| For Use With (Equipment) | E-Base™ |
| Gel Compatibility | E-Gel™ 48, E-Gel™ 96 |
| Gel Size | E-Gel™ 96 (13.5 x 10.8 cm), E-Gel™ 48 (13.5 x 10.8 cm) |
| Type | E-Holder Platform |
| Quantity | 2 units |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.