Learn More
Invitrogen™ Dynabeads™ Mouse DC (Dendritic Cell) Enrichment
Description
Includes
5mL Depletion MyOne; Streptavidin Dynabeads and 1mL Antibody Mix containing an optimized mixture of biotinylated monoclonal antibodies towards mouse CD2, CD3ε, CD49b, mIgM and Ter-119
The kit depletes T cells, mIgM+ B cells, NK cells, erythrocytes and most granulocytes. Highly enriched mouse DC's are bead and antibody-free, and can be used in any functional assay. The cells can be further sorted to obtain high purity of specific DC sub-populations. Obtain high recovery (80-100%) of viable Lin-CD11c+ DC's of all sup-populations, including DC's positive for CD4, CD8ε, B220 and CD19.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Type | Dendritic Cells |
| Content And Storage | The Kit Contains: 5 mL Depletion MyOne™ Streptavidin Dynabeads™ and 1 mL Antibody Mix containing an optimized mixture of biotinylated monoclonal antibodies towards mouse CD2, CD3ε, CD49b, mIgM and Ter-119. Store at 2-8°C |
| Product Type | Mouse DC Enrichment Kit |
| Isolation Technology | Negative Isolation |
| No. of Cells | Processes ∼1 x 109 cells total |
| Output Viability | >95% |
| Product Line | DYNAL, Dynabeads |
| Purity or Quality Grade | Research Grade |
| Quantity | 5 mL |
| Reactivity | Mouse |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.