missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Mini Tissue Kit
GREENER_CHOICE

Product Code. 10175572
Change view
Click to view available options
Quantity:
25 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175572 25 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10175572 Supplier Invitrogen™ Supplier No. CS11204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

  • ChargeSwitch Lysis Buffer (L13)
  • ChargeSwitch Lysis Buffer (L15)
  • ChargeSwitch Magnetic Beads
  • Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
  • RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
  • ChargeSwitch Purification Buffer (N5)
  • ChargeSwitch Wash Buffer (W12)
  • ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

The ChargeSwitch gDNA Mini Tissue Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from between 10-25 mg of tissue such as mouse tail, spleen, bone, and hair in less than 15 minutes after sample preparation and lysis.

The ChargeSwitch gDNA Mini Tissue Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from between 10-25 mg of tissue such as mouse tail, spleen, bone, and hair in less than 15 minutes after sample preparation and lysis.

  • Optimized for higher-purity DNA extraction
  • Improved performance with accompanying magnetic rack
  • Cleaner purified plasmid for optimal downstream performance
  • High Quality, High Yield Plasmid DNA
  • Compatible with MagnaRack™ Magnetic Rack
  • Improved Performance in Demanding Downstream Applications

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ gDNA Tissue Kits are shipped at room temperature. Upon receipt, store the Proteinase K and RNase A at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L13)
• ChargeSwitch™ Lysis Buffer (L15)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Scale Mini
Sample Type Tissue
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type gDNA Mini Tissue Kit
Quantity 25 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.