missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Mini Bacteria Kit

Product Code. 10319402
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10319402 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10319402 Supplier Invitrogen™ Supplier No. CS11301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
  • ChargeSwitch Resuspension Buffer (R4)
  • ChargeSwitch Lysis Buffer (L14)
  • ChargeSwitch Binding Buffer (B8)
  • ChargeSwitch Wash Buffer (W12)
  • ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
  • Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
  • RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)

PCR, restriction enzyme digestion, cloning, Southern blotting, and sequencing downstream applications

The ChargeSwitch gDNA Mini Bacteria Kit uses magnetic bead-based technology to provide rapid and efficient purification of up to 12μg of genomic DNA from up to 0.5mL of Gram-positive or Gram-negative bacterial culture or an individual colony in less than 15 minutes after lysate preparation

  • Optimized for higher-purity DNA extraction
  • Improved performance with accompanying magnetic rack
  • Cleaner purified plasmid for optimal downstream performance
  • High quality, high yield plasmid DNA
  • Compatible with MagnaRack™ magnetic rack
  • Improved performance in downstream applications
  • Less hazardous, less waste

DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Bacteria

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage All components of the ChargeSwitch™ gDNA (genomic DNA) Mini Bacteria Kit are shipped at room temperature. Upon receipt, store components as follows: Store Proteinase K at 4“C. Mix RNase A in Resuspension Buffer (R4) and store RNase A in Resuspension Buffer (R4) at 4“C. Store the remaining kit components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:

ChargeSwitch™ Magnetic Beads (25 mg/ml in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch™ Resuspension Buffer (R4)
ChargeSwitch™ Lysis Buffer (L14)
ChargeSwitch™ Binding Buffer (B8)
ChargeSwitch™ Wash Buffer (W12)
ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
Proteinase K (20 mg/ml in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
RNase A (5 mg/ml in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
Format Kit
Isolation Technology ChargeSwitch Chemistry
Scale Mini
Sample Type Bacteria
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type gDNA Mini Bacteria Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.