missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ INT 131

Description
INT 131 is a potent and selective PPARγ partial agonist (EC50 = 4 nM; Ki = 3.7 nM). It has a 20-fold higher affinity for PPARγ compared with that of Rosiglitazone (Cat. No. 5325). INT 131 improves glucose tolerance in a rodent model of diabetes without inducing hemodynamic and cardiovascular effects. Antidiabetic agent.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-N-[3,5-dichloro-4-(3-quinolinyloxy)phenyl]benzenesulfonamide |
| CAS | 315224-26-1 |
| Quantity | 50 mg |
| Target | PPAR gamma/NR1C3 |
| Molecular Formula | C21H12Cl4N2O3S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?