missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INSRR Polyclonal Antibody
GREENER_CHOICE

Product Code. 10101270
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10101270 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10101270 Supplier Invitrogen™ Supplier No. PA514691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

INSRR Polyclonal Antibody for Western Blot, IHC (P)

INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSRR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Insrr
Gene Accession No. P14616
Gene Alias Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr
Gene Symbols Insrr
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1256-1287 amino acids from the C-terminal region of human Insulin Receptor R.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.