Learn More
Invitrogen™ INSR alpha Monoclonal Antibody (83-7)

Description
This antibody is specific for IR and shows no cross-reactivity with insulin-like growth factor (IGF)-receptors. The epitope for this monoclonal antibody is conformational and is located in exon 3. Staining of formalin-fixed, paraffin tissues requires digestion of tissue sections with pepsin at 1mg/mL in Tris-HCl, pH 2.0, for 15 min at room temperature or 10 min at 37ºC. Recommended positive controls include IM-9 lymphocytes, placenta, or breast carcinoma.
Specifications
Specifications
| Antigen | INSR alpha |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 83-7 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | INSR |
| Gene Accession No. | P06213 |
| Gene Alias | 4932439J01Rik; CD220; CD220 beta; D630014A15Rik; DAF-2; HHF5; INSR; insulin receptor; insulin receptor (INSR); insulin receptor precursor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR alpha; IR beta; ir-A; IR-B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.