Learn More
Testo™ Immersion/Penetration Probe
Specifications
Specifications
| Type | Immersion/Penetration Probe |
| Certifications/Compliance | EN 60584-2 |
| Response Time | 5 sec. |
| Temperature Range (Metric) | -200°C to +1000°C |