missing translation for 'onlineSavingsMsg'
Learn More

JT Baker Imidazole BAKER ANALYZED Reagent 100g

Brand:  JT Baker 1747.0100


Imidazole BAKER ANALYZED Reagent 100g

Product Code. 10052350

  • ONLINE EXCLUSIVE PRICE
    £125.00
    £84.29 / 100g
    Save £40.71
    32% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.