missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL4 (Interleukin 4); Recombinant Human Protein

Product Code. 10161330
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10161330 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161330 Supplier Gibco™ Supplier No. CTP0043

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IL4 (Interleukin 4); Recombinant Human Protein

Recombinant IL4 is a bioactive protein intended for use in cell culture applications. IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors. ED50 = 0.05 to 0.4 ng⁄mL (Specific Activity: 2.0 x 10^7 to 2.5 x 10^6 units⁄mg). The biological activity is determined by measuring the dose-dependent proliferation of human TF-1 cells. A concentration range of 0.1 to 10.0 ng⁄mL is effective for most in vitro applications.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture, Primary Cell Culture, Stem Cell Culture
Molecular Weight (g/mol) 14.9 kDa
Name IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Purification Method Gel-purified
Quantity 1 mg
Storage Requirements 2°C to 8°C
Regulatory Status For research use only. Not for use in diagnostic procedures
Packaging 1 Ea.
Gene Alias IL-4 (Interleukin 4)
Common Name IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Product Type IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Specific Reactivity 0.187 ng/mL
Species Human
Expression System E. coli
Protein Family Cytokines and receptors
Protein Subtype Interleukins
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.