Learn More
Invitrogen™ IL-13 Polyclonal Antibody
Description
P130E targets IL-13 in ELISA, and WB applications and shows reactivity with Human samples. The P130E immunogen is recombinant human IL-13. P130E detects IL-13 which has a predicted molecular weight of approximately 13 kDa. The P130E IL13 polyclonal antibody has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M130B (biotinylated conjugate of clone 5A2) Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody P130E and biotinylated antibody M130B (clone 5A2) have successfully been used in combination with recombinant IL13 protein SIL13 in ELISA applications.
Specifications
Specifications
| Antigen | IL-13 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Il13 |
| Gene Accession No. | P35225 |
| Gene Alias | ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 |
| Gene Symbols | Il13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.