missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IHH Recombinant Rabbit Monoclonal Antibody (K01_2M85)
Description
IHH Recombinant Monoclonal Antibody for IHC (P), Western Blot
Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
Specifications
Specifications
| Antigen | IHH |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2M85 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | Ihh |
| Gene Accession No. | P97812, Q14623 |
| Gene Alias | BDA1; HHG2; HHG-2; Ihh; Indian hedgehog; Indian hedgehog homolog; indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product; indian hedgehog protein; LOW QUALITY PROTEIN: indian hedgehog protein; Indian hedgehog signaling molecule; Indian hedgehog-like protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?