missing translation for 'onlineSavingsMsg'
Learn More

IGFBP3 Polyclonal Antibody, Invitrogen™

Product Code. 10111140
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10111140 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10111140 Supplier Invitrogen Supplier No. PA514662

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and porcine based on sequence homology.

IGFBP3 is a gene that belongs to the insulin-like growth factor binding protein (IGFBP) family. The gene encodes a protein that contains both an IGFBP domain and a thyroglobulin type-I domain. This protein is known to form a ternary complex with insulin-like growth factor acid-labile subunit (IGFALS) and either insulin-like growth factor (IGF) I or II. The complex circulates in the plasma and prolongs the half-life of IGFs while also altering their interaction with cell surface receptors. Various isoforms of the gene have been identified through alternate transcriptional splice variants. Diseases associated with IGFBP3 include Insulin-Like Growth Factor I and Acid-Labile Subunit Deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene IGFBP3
Gene Accession No. P17936, P47878
Gene Alias acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703
Gene Symbols IGFBP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 162-189 amino acids from human IGFBP3
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100764858, 16009, 3486
Target Species Hamster, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.