missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma Monoclonal Antibody (A151D5B8), Invitrogen™

Product Code. 10120153
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10120153 500 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10120153 Supplier Invitrogen Supplier No. ASC4934

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN gamma
Applications ELISA
Classification Monoclonal
Clone A151D5B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene IFNG
Gene Accession No. P17803
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g
Gene Symbols IFNG
Host Species Mouse
Immunogen Recombinant swine IFN-gamma
Purification Method purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396991
Target Species Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.