missing translation for 'onlineSavingsMsg'
Learn More

Idebenone, MedChemExpress

Product Code. p-200099270
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16499046

Brand: Medchem Express HYN0303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Idebenone, a well-appreciated mitochondrial protectant, exhibits protective efficacy against neurotoxicity and can be used for the research of Alzheimer's disease, Huntington's disease.

Idebenone (oxidised form) has a dose-dependent inhibitory effect on the enzymatic metabolism of arachidonic acid in astroglial homogenates (IC50=16.65 μM). Idebenone, a coenzyme Q10 analog and an antioxidant, induces apoptotic cell death in the human dopaminergic neuroblastoma SHSY-5Y cells. Idebenone quickly crosses the blood-brain barrier.

Specifications

Chemical Name or Material Idebenone
CAS 58186-27-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H30O5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (295.47 mM)
SMILES O=C1C(CCCCCCCCCCO)=C(C)C(C(OC)=C1OC)=O
Molecular Weight (g/mol) 338.44
Formula Weight 338.44
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.