missing translation for 'onlineSavingsMsg'
Learn More

IAXO-102, MedChemExpress

Product Code. 16455906
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455906 100 mg 100mg
16431447 50 mg 50mg
16407957 5 mg 5mg
16405238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16455906 Supplier Medchem Express Supplier No. HY125171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. IAXO-102 inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development.

IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. IAXO-102 inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development.

Specifications

Chemical Name or Material IAXO-102
Physical Form Solid
CAS 1115270-63-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C35H71NO5
For Use With (Application) COVID-19-immunoregulation
Solubility Information Ethanol : 50 mg/mL (85.33 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.53 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCO[C@@H]1[C@H]([C@@H]([C@@H](CN)O[C@@H]1OC)O)OCCCCCCCCCCCCCC
Molecular Weight (g/mol) 585.94
Formula Weight 585.94
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.